Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD4 Antibody (RAB-C131), Biotin - Chimeric, Novus Biologicals™
SDP

Catalog No. NB234580 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB234580 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB234580 Supplier Novus Biologicals Supplier No. NBP320125B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BRD4 Monoclonal antibody specifically detects BRD4 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen BRD4
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone RAB-C131
Conjugate Biotin
Formulation PBS
Gene Alias bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing BRD4 protein under non-denaturing conditions.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23476
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.