Learn More
BRD4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$568.26 - $977.24
Specifications
| Antigen | BRD4 |
|---|---|
| Concentration | 1 mg/ml |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-1488
|
Bethyl Laboratories, Inc
A301985A50 |
50 μL |
Each for $568.26
|
|
|||||
50-156-1487
|
Bethyl Laboratories, Inc
A301985A100 |
100 μL |
Each for $977.24
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains.BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.Specifications
| BRD4 | |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O60885, Q9ESU6 | |
| 23476, 57261 | |
| Between 1312 and 1362 | |
| Primary | |
| 4° C | |
| BRD4 |
| 1 mg/ml | |
| Polyclonal | |
| Liquid | |
| RUO | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| HUNK1 | |
| Brd4 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.