Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD4 Rabbit anti-Human, Mouse, Rat, Clone: 3V5R1, Novus Biologicals™
SDP

Catalog No. NB168994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168994 20 μg
NB168993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168994 Supplier Novus Biologicals Supplier No. NBP31695820UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BRD4 Monoclonal antibody specifically detects BRD4 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen BRD4
Applications Western Blot
Classification Monoclonal
Clone 3V5R1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T204 of human BRD4 (NP_490597.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23476
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.