Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRD4 Recombinant Rabbit Monoclonal Antibody (BL-149-2H5)

Rabbit Recombinant Monoclonal Antibody
$173.10 - $458.00
Specifications
| Antigen | BRD4 |
|---|---|
| Clone | BL-149-2H5 |
| Concentration | 100 μg/mL |
| Content And Storage | 4°C |
| Applications | ChIP sequencing (ChIP-seq), Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544203
![]() |
Invitrogen™
MA544203 |
100 μL |
Each for $458.00
|
|
|||||
|
PIMA544204
|
Invitrogen™
MA544204 |
20 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE tissue sections.
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.Specifications
| BRD4 | |
| 100 μg/mL | |
| ChIP sequencing (ChIP-seq), Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
| O60885 | |
| 23476 | |
| Synthetic peptide representing a region between residues 1312 and 1362 of human bromodomain-containing protein 4. | |
| Antibody |
| BL-149-2H5 | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| BRD4 | |
| Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; bromodomain-containing protein 4; bromodomain-containing protein 4; LOW QUALITY PROTEIN: bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; mitotic chromosome-associated protein; Protein HUNK1; WI-11513 | |
| BRD4 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title