Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRD4 Recombinant Rabbit Monoclonal Antibody (BL-151-6F11)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | BRD4 |
---|---|
Clone | BL-151-6F11 |
Concentration | 100 μg/mL |
Content And Storage | 4°C |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544205
|
Invitrogen™
MA544205 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544206
|
Invitrogen™
MA544206 |
20 μL |
Each for $173.10
|
|
|||||
Description
BRD4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, IP
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.Specifications
BRD4 | |
100 μg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
O60885, Q9ESU6 | |
23476, 57261 | |
Synthetic peptide representing a region between residues 1312 and 1362 of human bromodomain-containing protein 4. | |
Antibody |
BL-151-6F11 | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
BRD4 | |
Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; bromodomain-containing protein 4; bromodomain-containing protein 4; LOW QUALITY PROTEIN: bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; mitotic chromosome-associated protein; Protein HUNK1; WI-11513 | |
BRD4 | |
Primary | |
purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title