Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP128727 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128727 Supplier Novus Biologicals Supplier No. NBP128727
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRD7 Polyclonal specifically detects BRD7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).

Specifications

Antigen BRD7
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2 -10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q9NPI1
Gene Alias 75 kDa bromodomain protein, BP75bromodomain-containing 7, bromodomain containing 7, bromodomain-containing protein 7, CELTIX1NAG4, Protein CELTIX-1
Gene Symbols BRD7
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 601 and 651 of human bromodomain containing 7 using the numbering given in entry NP_037395.2 (GeneID 29117).
Molecular Weight of Antigen 74 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29117
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.