Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD8 Antibody, Novus Biologicals™
SDP

Catalog No. NB10079781 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10079781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10079781 Supplier Novus Biologicals Supplier No. NB10079781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRD8 Polyclonal specifically detects BRD8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen BRD8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:25000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-8 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q969M9
Gene Alias bromodomain containing 8, bromodomain-containing protein 8, Skeletal muscle abundant protein, SMAP2, SMAPp120Skeletal muscle abundant protein 2, Thyroid hormone receptor coactivating protein of 120 kDa, trCP120
Gene Symbols BRD8
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 200 and 250 of human Bromodomain containing 8 using the numbering given in TrEMBL entry Q969M9 (GeneID 10902).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 10902
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.