Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRD9 Recombinant Rabbit Monoclonal Antibody (BLR153J)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | BRD9 |
---|---|
Clone | BLR153J |
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544491
|
Invitrogen™
MA544491 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544492
|
Invitrogen™
MA544492 |
10 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
Bromodomain containing protein 9 (BRD9) is an uncharacterized protein that contains one bromodomain.Specifications
BRD9 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
Q9H8M2 | |
65980 | |
Peptide representing a region between residue 547 and the C-terminus (residue 597) of Human Bromodomain-containing protein 9. | |
Antibody |
BLR153J | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
BRD9 | |
AL022779; Brd9; bromodomain containing 9; bromodomain-containing protein 9; DKFZp434D0711; DKFZp686L0539; FLJ43530; LAVS3040; PRO9856; rhabdomyosarcoma antigen MU-RMS-40.8; sarcoma antigen NY-SAR-29; UNQ3040/PRO9856 | |
BRD9 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title