Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Breast cancer suppressor candidate 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215615 Supplier Novus Biologicals Supplier No. NBP215615
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Breast cancer suppressor candidate 1 Polyclonal specifically detects Breast cancer suppressor candidate 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Breast cancer suppressor candidate 1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O00534
Gene Alias BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein, BCSC1ortholog of mouse AW551984, Breast cancer suppressor candidate 1, LOH11CR2Avon Willebrand factor A domain-containing protein 5A, loss of heterozygosity, 11, chromosomal region 2, gene A, von Willebrand factor A domain containing 5A
Gene Symbols VWA5A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human VWA5A. The exact sequence is proprietary.
Molecular Weight of Antigen 86 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4013
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.