Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Breast cancer suppressor candidate 1 Antibody (OTI3D6), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27118A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27118A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27118A32 Supplier Novus Biologicals Supplier No. NBP271158AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Breast cancer suppressor candidate 1 Monoclonal specifically detects Breast cancer suppressor candidate 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Breast cancer suppressor candidate 1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3D6
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein, BCSC1ortholog of mouse AW551984, Breast cancer suppressor candidate 1, LOH11CR2Avon Willebrand factor A domain-containing protein 5A, loss of heterozygosity, 11, chromosomal region 2, gene A, von Willebrand factor A domain containing 5A
Gene Symbols VWA5A
Host Species Mouse
Immunogen Full length recombinant protein of human Breast cancer suppressor candidate 1 (NP_055437) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4013
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.