Learn More
Breast cancer suppressor candidate 1 Antibody (OTI3D6), PE/Cy5.5, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Breast cancer suppressor candidate 1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI3D6 |
| Conjugate | PE-Cyanine5.5 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein, BCSC1ortholog of mouse AW551984, Breast cancer suppressor candidate 1, LOH11CR2Avon Willebrand factor A domain-containing protein 5A, loss of heterozygosity, 11, chromosomal region 2, gene A, von Willebrand factor A domain containing 5A |
| Gene Symbols | VWA5A |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.