Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Breast carcinoma amplified sequence 3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB105888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105888 Supplier Novus Biologicals Supplier No. NBP298807AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Breast carcinoma amplified sequence 3 Polyclonal specifically detects Breast carcinoma amplified sequence 3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Breast carcinoma amplified sequence 3
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias BCAS4/BCAS3 fusion, breast carcinoma amplified sequence 3, breast carcinoma amplified sequence 4/3 fusion protein, breast carcinoma-amplified sequence 3, DKFZp686O1527, FLJ20128, GAOB1, MAAB, metastasis associated antigen of breast cancer, MGC4973, protein Maab1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Breast carcinoma amplified sequence 3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54828
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.