Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Brefeldin A
GSA_VAGREENER_CHOICESDP

Catalog No. 12315 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-315 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-315 Supplier R&D Systems™ Supplier No. 1231/5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Disrupts protein translocation to Golgi

Reversible inhibitor of protein translocation from the endoplasmic reticulum (ER) to the Golgi apparatus. Blocks binding of ADP-ribosylation factor (ARF1) to the Golgi apparatus and inhibits GDP-GTP exchange. Can be used to induce autophagy in mammalian cells. Also enhances CRISPR-mediated homology-directed repair (HDR) efficiency ∼2-fold when applied at 100 nM, in human induced pluripotent stem cells (iPSCs). Antifungal.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 20350-15-6
Molecular Formula C16H24O4

Specifications

Quantity 5 mg
Product Line Tocris
Formula Weight 280.36
Purity >98%
Molecular Weight (g/mol) 280.36
Chemical Name or Material 1,6,7,8,9,11 Aβ,12,13,14,14 Aα-Decahydro-1β,13α-dihydroxy-6β-methyl-4 H-cyclopent[f]oxacyclotridecin-4-one
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.