Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brentuximab Antibody (2262C), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345668 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345668 Supplier Novus Biologicals Supplier No. FAB9584MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Brentuximab Monoclonal specifically detects Brentuximab in Multi-species samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen Brentuximab
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2262C
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Full length Brentuximab
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.