Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Brequinar sodium
GREENER_CHOICE

Catalog No. p-7114675
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective DHODH inhibitor.

Brequinar sodium is a potent and selective dihydroorotate dehydrogenase (DHODH) inhibitor (IC50 = ∽20 nM). Causes depletion of pyrimidine nucleotides. Exhibits minimal inhibition against a panel of >400 kinases. Triggers differentiation of AML cell lines (ED50 = ∽ 1 μM. Note uridine concentration effects ED50 value). Causes differentiation and depletion of leukemia-initiating cells in vivo. Antimetabolite.

Specifications

Chemical Name or Material 6-Fluoro-2-(2′-fluoro[1,1′-biphenyl]-4-yl)-3-methyl-4-quinolinecarboxylic acid sodium
CAS 96201-88-6
Assay Percent Range >99%
Quantity 10 mg
Target DHODH Inhibitors
Molecular Formula C23H14F2NO2Na
Formula Weight Observed MW: 397.35
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.