Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brevican Antibody (S294A-6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB222401647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222401647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222401647 Supplier Novus Biologicals Supplier No. NBP222401AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Brevican Monoclonal specifically detects Brevican in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Brevican
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S294A-6
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BEHABbrevican core protein, Brain-enriched hyaluronan-binding protein, brevican, Chondroitin sulfate proteoglycan 7, chondroitin sulfate proteoglycan BEHAB, chondroitin sulfate proteoglycan BEHAB/brevican, CSPG7brevican proteoglycan, MGC13038
Gene Symbols BCAN
Host Species Mouse
Immunogen Fusion protein amino acids 219-655 of rat Brevican
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 63827
Test Specificity Detects approx 140kDa (and smaller due to proteolytic cleavage).
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.