Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRF1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP18044020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18044020 20 μL
NBP180440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18044020 Supplier Novus Biologicals Supplier No. NBP18044020UL

Rabbit Polyclonal Antibody

BRF1 Polyclonal specifically detects BRF1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen BRF1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_082469
Gene Alias B-related factor 1, BRF-1, BRF1 homolog, subunit of RNA polymerase III transcription initiation factorIIIB (S. cerevisiae), BRFFLJ42674, general transcription factor IIIB, 90kD subunit, GTF3BTAFIII90, hBRFMGC105048, hTFIIIB90, TAF3B2, TAF3CB - related factor 1, TATA box binding protein (TBP)-associated factor 3C, TATA box binding protein (TBP)-associated factor, RNA polymerase III, GTF3Bsubunit 2, TATA box binding protein (TBP)-associated factor, RNA polymerase III, subunit 2, TATA box-binding protein-associated factor, RNA polymerase III, subunit 2, TBP - associated factor, RNA polymerase III, 90kD, TF3B90, TFIIIB90FLJ43034, transcription factor IIIB 90 kDa subunit
Gene Symbols BRF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal of mouse BRF1. Peptide sequence GSPPRDDSQPPERASTKKLSRRKRATTRNSADPGTSTGKRLRPLLSAQPA.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2972
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.