Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | BRF1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18044020
![]() |
Novus Biologicals
NBP18044020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180440
![]() |
Novus Biologicals
NBP180440 |
100 μL |
Each for $487.50
|
|
|||||
Description
BRF1 Polyclonal specifically detects BRF1 in Human samples. It is validated for Western Blot.Specifications
BRF1 | |
Polyclonal | |
Rabbit | |
NP_082469 | |
2972 | |
Synthetic peptide directed towards the C terminal of mouse BRF1. Peptide sequence GSPPRDDSQPPERASTKKLSRRKRATTRNSADPGTSTGKRLRPLLSAQPA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
B-related factor 1, BRF-1, BRF1 homolog, subunit of RNA polymerase III transcription initiation factorIIIB (S. cerevisiae), BRFFLJ42674, general transcription factor IIIB, 90kD subunit, GTF3BTAFIII90, hBRFMGC105048, hTFIIIB90, TAF3B2, TAF3CB - related factor 1, TATA box binding protein (TBP)-associated factor 3C, TATA box binding protein (TBP)-associated factor, RNA polymerase III, GTF3Bsubunit 2, TATA box binding protein (TBP)-associated factor, RNA polymerase III, subunit 2, TATA box-binding protein-associated factor, RNA polymerase III, subunit 2, TBP - associated factor, RNA polymerase III, 90kD, TF3B90, TFIIIB90FLJ43034, transcription factor IIIB 90 kDa subunit | |
BRF1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title