Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRF2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001120 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001120 Supplier Novus Biologicals Supplier No. NB1001120
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BRF2 Polyclonal specifically detects BRF2 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen BRF2
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_060780.
Gene Alias B-related factor 2, BRF-2, BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like, BRFUhTFIIIB50, FLJ11052, hBRFU, TFIIIB50RNA polymerase III transcription initiation factor BRFU, transcription factor IIB- related factor, TFIIIB50, transcription factor IIIB 50 kDa subunit
Gene Symbols BRF2
Host Species Goat
Immunogen Peptide with sequence C-QAARQAATSVPNPP corresponding to C-Terminus according to NP_060780.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55290
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.