Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brg1 Antibody (22F3), Novus Biologicals™
SDP

Catalog No. NB298348 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298348 50 μL
NB298349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298348 Supplier Novus Biologicals Supplier No. NBP32631450UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Brg1 Monoclonal antibody specifically detects Brg1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen Brg1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 22F3
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias ATP-dependent helicase SMARCA4, BAF190, BAF190A, brahma protein-like 1, BRG1-associated factor 190A, BRG1SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 4, BRM/SWI2-related gene 1, EC 3.6.1, EC 3.6.4.-, FLJ39786, global transcription activator homologous sequence, homeotic gene regulator, hSNF2b, Mitotic growth and transcription activator, nuclear protein GRB1, Protein brahma homolog 1, Protein BRG-1, RTPS2, SNF2, SNF2B, SNF2-BETA, SNF2L4, SNF2LB, SNF2-like 4, subfamily a, member 4, sucrose nonfermenting-like 4, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI2, transcription activator BRG1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Brg1 [UniProt P51532]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Modifiers, Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6597
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.