Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRI3BP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | BRI3BP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP162385
![]() |
Novus Biologicals
NBP162385 |
100 μL |
Each for $487.50
|
|
|||||
NBP16238520
![]() |
Novus Biologicals
NBP16238520UL |
20 μL | N/A | N/A | N/A | ||||
Description
BRI3BP Polyclonal specifically detects BRI3BP in Human samples. It is validated for Western Blot.Specifications
BRI3BP | |
Polyclonal | |
Rabbit | |
Q8WY22 | |
140707 | |
Synthetic peptides corresponding to BRI3BP(BRI3 binding protein) The peptide sequence was selected from the C terminal of BRI3BP. Peptide sequence GFYWRSSPSGPSNPSNPSVEEKLEHLEKQVRLLNIRLNRVLESLDRSKDK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
BNAS1, BRI3 binding protein, BRI3-binding protein, Cervical cancer 1 proto-oncogene-binding protein KG19, cervical cancer oncogene binding protein, HCCR-1, HCCR-2, HCCRBP-1, KG19I3-binding protein | |
BRI3BP | |
IgG | |
28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title