Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij(R) S100, Spectrum™ Chemical
SDP

Catalog No. 18601070 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail

B1593, 9005-00-9

Spectrum™ Chemical Brij(R) S100 , Polyethylene (100) Stearyl Ether, functions as an aqueous dispersant and oil/water emulsifier.

Chemical Identifiers

CAS 9005-00-9
Molecular Formula C20H41O2
Molecular Weight (g/mol) 313.55
InChI Key PSEGVHKUPXNGGJ-UHFFFAOYSA-N
IUPAC Name [2-(octadecyloxy)ethyl]oxidanyl
SMILES CCCCCCCCCCCCCCCCCCOCC[O]

Specifications

CAS Max % 100%
Quantity 500 g
Packaging Amber Glass Bottle
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.