Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRIP1/FANCJ Antibody (pp15-IB4), Novus Biologicals™
SDP

Catalog No. NB100189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100189 0.1 mL
NB100189SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100189 Supplier Novus Biologicals Supplier No. NB100189
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

BRIP1/FANCJ Monoclonal specifically detects BRIP1/FANCJ in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen BRIP1/FANCJ
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone pp15-IB4
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 1:10-1:500
Formulation Ascites with 0.05% Sodium Azide
Gene Accession No. Q9BX63
Gene Alias ATP-dependent RNA helicase BRIP1, BACH1BRCA1/BRCA2-associated helicase 1, BRCA1 interacting protein C-terminal helicase 1, BRCA1-associated C-terminal helicase 1, BRCA1-binding helicase-like protein BACH1, BRCA1-interacting protein 1, BRCA1-interacting protein C-terminal helicase 1, EC 3.6.1, EC 3.6.4.13, FANCJFanconi anemia group J protein, FLJ90232, MGC126521, MGC126523, OF, Protein FACJ
Gene Symbols BRIP1
Host Species Mouse
Immunogen A peptide corresponding to the C-terminus of human FANCJ (within amino acids 1200-1249). [SwissProt# Q9BX63]
Molecular Weight of Antigen 130 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 83990
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.