Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRIP1/FANCJ Antibody (SR1035), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322189100UL
Description
BRIP1/FANCJ Monoclonal antibody specifically detects BRIP1/FANCJ in Human samples. It is validated for Western BlotSpecifications
BRIP1/FANCJ | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1035 | |
Western Blot 1:500-1:2000 | |
ATP-dependent RNA helicase BRIP1, BACH1BRCA1/BRCA2-associated helicase 1, BRCA1 interacting protein C-terminal helicase 1, BRCA1-associated C-terminal helicase 1, BRCA1-binding helicase-like protein BACH1, BRCA1-interacting protein 1, BRCA1-interacting protein C-terminal helicase 1, EC 3.6.1, EC 3.6.4.13, FANCJFanconi anemia group J protein, FLJ90232, MGC126521, MGC126523, OF, Protein FACJ | |
A synthesized peptide derived from human BRIP1/FANCJ (Uniprot #: Q9BX63) | |
100 μg | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
83990 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction