Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRIP1/FANCJ Antibody (SR1035), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | BRIP1/FANCJ |
---|---|
Clone | SR1035 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
BRIP1/FANCJ Monoclonal antibody specifically detects BRIP1/FANCJ in Human samples. It is validated for Western BlotSpecifications
BRIP1/FANCJ | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ATP-dependent RNA helicase BRIP1, BACH1BRCA1/BRCA2-associated helicase 1, BRCA1 interacting protein C-terminal helicase 1, BRCA1-associated C-terminal helicase 1, BRCA1-binding helicase-like protein BACH1, BRCA1-interacting protein 1, BRCA1-interacting protein C-terminal helicase 1, EC 3.6.1, EC 3.6.4.13, FANCJFanconi anemia group J protein, FLJ90232, MGC126521, MGC126523, OF, Protein FACJ | |
A synthesized peptide derived from human BRIP1/FANCJ (Uniprot #: Q9BX63) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1035 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
83990 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title