Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRIP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501557924
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
Catalog No. Quantity
50-155-7924 10 μL
1 options

Catalog No. 50-155-7924

Supplier: Bethyl Laboratories, Inc A300560AT

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The protein encoded by this gene is a member of the RecQ DEAH helicase family and interacts with the BRCT repeats of breast cancer, type 1 (BRCA1). The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). This gene may be a target of germline cancer-inducing mutations.

Specifications

Antigen BRIP1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene BRIP1
Gene Accession No. Q9BX63
Gene Alias FANCJ protein, BACH 1, BACH1 BRIP1, BRIP 1, BRAC 1 Associated C Terminal Helicase 1, BRCA 1 Interacting Protein 1
Gene Symbols BRIP1
Host Species Rabbit
Immunogen Between 1100 and 1150
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83990
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.