Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRIX Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB084494 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084494 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084494 Supplier Novus Biologicals Supplier No. NBP299257AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRIX Polyclonal specifically detects BRIX in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BRIX
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias brix domain containing 2, Brix domain-containing protein 2, BRIXBXDC2, BRX1, biogenesis of ribosomes, homolog (S. cerevisiae), FLJ11100, ribosome biogenesis protein BRX1 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human BRIX.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 55299
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.