Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brk Antibody (2H12B8), Novus Biologicals™
SDP

Catalog No. NB11060537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060537 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060537 Supplier Novus Biologicals Supplier No. NB11060537
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Brk Monoclonal specifically detects Brk in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Brk
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2H12B8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias Breast tumor kinase, BRKprotein-tyrosine kinase 6, EC 2.7.10, EC 2.7.10.2, FLJ42088, protein-tyrosine kinase BRK, PTK6 protein tyrosine kinase 6, Tyrosine-protein kinase BRK
Gene Symbols PTK6
Host Species Mouse
Immunogen Ni-NTA purified truncated recombinant PTK6 expressed in E. Coli strain BL21 (DE3).
Molecular Weight of Antigen 52 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5753
Test Specificity PTK6 (2H12B8)
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b Kappa
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.