Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brk Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB082067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082067 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082067 Supplier Novus Biologicals Supplier No. NBP298347FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Brk Polyclonal specifically detects Brk in Mouse samples. It is validated for ELISA.

Specifications

Antigen Brk
Applications ELISA
Classification Polyclonal
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias Breast tumor kinase, BRKprotein-tyrosine kinase 6, EC 2.7.10, EC 2.7.10.2, FLJ42088, protein-tyrosine kinase BRK, PTK6 protein tyrosine kinase 6, Tyrosine-protein kinase BRK
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Brk (Q64434; Met1-Val451).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5753
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.