Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BRL 50481

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, substrate-competitive inhibitor of phosphodiesterase (PDE) 7 (Ki = 180 nM). Displays > 200-fold selectivity over PDE1B, PDE1C, PDE2, PDE3, PDE4A4 and PDE5.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 244.26 |
| Purity | >99% |
| Molecular Weight (g/mol) | 244.265 |
| Chemical Name or Material | BRL 50481 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction