Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRMS1 Mouse anti-Human, Unconjugated, Clone: 3a1.21, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | BRMS1 |
---|---|
Clone | 3a1.21 |
Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC17532MI
|
MilliporeSigma
MABC175325UL |
25 μL |
Each for $189.29
|
|
|||||
MABC17531MI
|
MilliporeSigma
MABC1753100UL |
100 μL |
Each for $477.80
|
|
|||||
Description
BRMS1 Monoclonal antibody specifically detects BRMS1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting.Specifications
BRMS1 | |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Breast cancer metastasis-suppressor 1 | |
GST-tagged full-length recombinant human Breast cancer metastasis-suppressor 1. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼35 kDa observed; 28.46 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
3a1.21 | |
Monoclonal | |
Purified | |
Apoptosis and Cancer | |
NP_056214 | |
BRMS1 | |
IgG1 κ | |
Protein G Purified | |
Clone 3a1.21 is a mouse monoclonal antibody that detects human Breast cancer metastasis-suppressor 1. It targets an epitope within the first 50 amino acids from the N-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title