Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRN3B/POU4F2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP226061 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226061 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226061 Supplier Novus Biologicals Supplier No. NBP226061
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BRN3B/POU4F2 Polyclonal specifically detects BRN3B/POU4F2 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen BRN3B/POU4F2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias POU domain, class 4, transcription factor 2, BRN3.2, Brn-3b, BRN3B, POU class 4 homeobox 2
Gene Symbols POU4F2
Host Species Goat
Immunogen Peptide with sequence HGLPSHMGCMSDVD corresponding to internal region according to NP_004566.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5458
Test Specificity This antibody is not expected to cross-react to the similar POU4F3 (GeneID 5459)
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.