Learn More
Bromoacetaldehyde dimethyl acetal, 97+%, stab. with potassium carbonate
Description
2-Bromo-1,1-dimethoxyethane (Bromoacetaldehyde dimethyl acetal) was used in the synthesis of 2,3-O-acetal via reaction with 2,3-diol in the presence of 10-camphorsulphonic acid (CSA). 2-bromo-1,1-dimethoxyethane was used to react with 4-hydroxylbenzaldehyde, and finally aldehyde substituted prodrug was generated by TFA-mediated deacetalization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromo-1,1-dimethoxyethane (Bromoacetaldehyde dimethyl acetal) was used in the synthesis of 2,3-O-acetal via reaction with 2,3-diol in the presence of 10-camphorsulphonic acid (CSA). 2-bromo-1,1-dimethoxyethane was used to react with 4-hydroxylbenzaldehyde, and finally aldehyde substituted prodrug was generated by TFA-mediated deacetalization.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
Specifications
Specifications
| Density | 1.435 |
| Boiling Point | 150°C |
| Flash Point | 54°C (129°F) |
| Refractive Index | 1.445 |
| Linear Formula | BrCH2CH(OCH3)2 |
| Quantity | 1000 g |
| UN Number | UN2929 |
| Beilstein | 1280132 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H301-H315-H319-H330-H335 |
| Hazard Statement | GHS H Statement H301-H330-H226-H315-H319-H335 Toxic if swallowed. Fatal if inhaled. Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S. |
| EINECSNumber | 230-669-6 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.