Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodeoxyuridine/BrdU Antibody (BU-1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB302788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302788 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302788 Supplier Novus Biologicals Supplier No. FAB7225N

Mouse Monoclonal Antibody

Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Broad Species Reactivity samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Bromodeoxyuridine/BrdU
Applications Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone BU-1
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Bromodeoxyuridine, BUdr
Host Species Mouse
Immunogen 5-iodourindine (5-IO) coupled to ovalbumin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.