Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodeoxyuridine/BrdU Antibody (BU-1), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346623 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346623 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346623 Supplier Novus Biologicals Supplier No. FAB7225MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bromodeoxyuridine/BrdU Monoclonal specifically detects Bromodeoxyuridine/BrdU in Multi-species samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry.

Specifications

Antigen Bromodeoxyuridine/BrdU
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone BU-1
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Bromodeoxyuridine, BUdr
Host Species Mouse
Immunogen 5-iodourindine (5-IO) coupled to ovalbumin
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.