Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodeoxyuridine/BrdU Antibody (MoBu-1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB008609 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008609 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008609 Supplier Novus Biologicals Supplier No. NBP268041APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bromodeoxyuridine/BrdU Monoclonal specifically detects Bromodeoxyuridine/BrdU in All Species samples. It is validated for Flow Cytometry.

Specifications

Antigen Bromodeoxyuridine/BrdU
Applications Flow Cytometry
Classification Monoclonal
Clone MoBu-1
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Bromodeoxyuridine, BUdr
Host Species Mouse
Immunogen Bromodeoxyuridine (BrdU) conjugated to hemocyanine (isolated from Helix pomatia)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity It reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.
Target Species All species
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.