Learn More
Bromodeoxyuridine/BrdU Antibody (SPM537), DyLight 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Bromodeoxyuridine/BrdU |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | SPM537 |
| Conjugate | DyLight 405 |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | Bromodeoxyuridine, BUdr |
| Host Species | Mouse |
| Immunogen | Bromodeoxyuridine (BrdU) conjugated to KLH |
| Purification Method | Protein A or G purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.