Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BromoTag Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB335274 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335274 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335274 Supplier Novus Biologicals Supplier No. NBP317999MFV450
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

BromoTag Polyclonal antibody specifically detects BromoTag in Nonspecies specific samples. It is validated for Western Blot

Specifications

Antigen BromoTag
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Brd4 bromodomain L387A variant, Brd4BD2L387A, BromoTag degron
Host Species Sheep
Immunogen Partial recombinant protein made to the second bromodomain of Brd4 with a L387A mutation (see Bond, et al PMID 34652918).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.