Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromotris(dimethylamino)phosphonium hexafluorophosphate, 98%, Thermo Scientific™

Catalog No. AC456700050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456700050 5 g
AC456700010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456700050 Supplier Thermo Scientific Chemicals Supplier No. 456700050

Chemical Identifiers

CAS 50296-37-2
Molecular Formula C6H18BrF6N3P2
Molecular Weight (g/mol) 388.07
MDL Number MFCD00191864
InChI Key XELPBWPBGHCIKX-UHFFFAOYSA-N
Synonym bromotris dimethylamino phosphonium hexafluorophosphate, brop, bromotris dimethylamino phosphonium hexafluorophosphate v, bromo-tris dimethylamino phosphonium hexafluorophosphate, acmc-20aljt, ksc269e9j, bromotris dimethylamino phosphonium hexafluorophos, bromotris dimethylamino phosphoniumhexafluorophosphate, bromanyl-tris dimethylamino phosphanium hexafluorophosphate
PubChem CID 10221847
IUPAC Name bromo-tris(dimethylamino)phosphanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CN(C)[P+](Br)(N(C)C)N(C)C

Specifications

Melting Point 249.0°C to 252.0°C
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 388.07
Percent Purity 98%
Chemical Name or Material Bromotris(dimethylamino)phosphonium hexafluorophosphate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage. May cause cancer by inhalation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.