Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brompheniramine Maleate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601108 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-108 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-108 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BR16725GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

BR167, 980-71-2, C16H19BrN2·C4H4O4

Spectrum™ Chemical Brompheniramine Maleate, USP is an antihistamine used to alleviate symptoms of the common cold and allergic rhinitis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 980-71-2
Molecular Formula C20H23BrN2O4
Molecular Weight (g/mol) 435.32
MDL Number MFCD00057367
InChI Key SRGKFVAASLQVBO-BTJKTKAUNA-N
IUPAC Name (2Z)-but-2-enedioic acid; [3-(4-bromophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
SMILES OC(=O)\C=C/C(O)=O.CN(C)CCC(C1=CC=C(Br)C=C1)C1=CC=CC=N1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +0.2° to -0.2°
Percent Purity 98-102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.