Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brorin/VWC2 Antibody (08), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB328621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328621 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328621 Supplier Novus Biologicals Supplier No. NBP305797MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Brorin/VWC2 Monoclonal antibody specifically detects Brorin/VWC2 in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen Brorin/VWC2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 08
Conjugate mFluor Violet 500 SE
Dilution ELISA, Sandwich ELISA Capture
Formulation 50mM Sodium Borate
Gene Alias Brain-Specific Chordin-Like, Brain-Specific Chordin-Like Protein, brorin, PSST739, UNQ739, Von Willebrand Factor C Domain Containing 2, Von Willebrand Factor C Domain-Containing Protein 2
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Brorin/VWC2 (Accession#: NP_940972.2; Met1-Met325).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375567
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.