Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Brownlee Electrolysis Apparatus
SDPSCI_ED

Catalog No. S139458 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Activity Guide, Platinum Electrode Assembly, Two Glass Test Tubes
Beaker, Activity Guide, Platinum Electrode Assembly, Two Glass Test Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S139458 Beaker, Activity Guide, Platinum Electrode Assembly, Two Glass Test Tubes
S05931 Activity Guide, Platinum Electrode Assembly, Two Glass Test Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S139458 Supplier United Scientific™ Supplier No. BEA001B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Brownlee Electrolysis Apparatus is a simple and economical type of electrolysis apparatus.

  • Platinum electrodes are attached to insulated connecting rods, which are attached to binding posts mounted on a non-conducting support.
  • Support rests across the top of a battery jar (not included in both sets).
  • Support comes equipped with two clips, each holding an inverted test tube.
  • Unit operates on a user supplied 6 V battery or 10 V DC power supply.

Specifications

Type Electrolysis Apparatus
Includes Beaker, Activity Guide, Platinum Electrode Assembly, Two Glass Test Tubes
Power Supply 6 V Battery or 10 VDC Power Supply
Material Borosilicate Glass
Quantity 1 Ea.
For Use With (Application) Useful device for the study of electrolysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.