Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRPF1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215620 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215620 Supplier Novus Biologicals Supplier No. NBP215620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRPF1 Polyclonal antibody specifically detects BRPF1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin).

Specifications

Antigen BRPF1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated
Formulation PBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300
Gene Accession No. P55201
Gene Alias bromodomain and PHD finger containing, 1, Bromodomain and PHD finger-containing protein 1, peregrin, Protein Br140
Gene Symbols BRPF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human BRPF1. The exact sequence is proprietary.
Molecular Weight of Antigen 137 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7862
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.