Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRSK2 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB218330 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB218330 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB218330 Supplier Novus Biologicals Supplier No. NBP177281IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRSK2 Polyclonal antibody specifically detects BRSK2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen BRSK2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias BR serine/threonine kinase 2, BR serine/threonine-protein kinase 2, C11orf7, chromsosome 11 open reading frame 7, EC 2.7.11, EC 2.7.11.1, FLJ41362, PEN11BSAD1, protein kinase SAD1B, serine/threonine kinase 29, serine/threonine kinase SAD-A, Serine/threonine-protein kinase 29, Serine/threonine-protein kinase SAD-A, STK29
Host Species Rabbit
Immunogen Antibody was raised against a 23 amino acid synthetic peptide from near the carboxy terminus of human BRSK2. The immunogen is located within amino acids 630 - 680 of BRSK2. Amino Acid Squence: DPPAAQHLSDTTNNCMEMMTGRLS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9024
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.