Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brucella Abortus Antibody (BrG11), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10073111R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073111R 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073111R Supplier Novus Biologicals Supplier No. NB10073111R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Brucella Abortus Monoclonal specifically detects Brucella Abortus in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Brucella Abortus
Applications ELISA, Western Blot
Classification Monoclonal
Clone BrG11
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B.abortus
Host Species Mouse
Immunogen This is reacting with intact Brucella-cells.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity LPS of Brucella abortus and intact Brucella-cells.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.