Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRUNOL4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$382.00 - $646.00
Specifications
Antigen | BRUNOL4 |
---|---|
Concentration | 0.2mg/mL |
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Description
BRUNOL4 Polyclonal specifically detects BRUNOL4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BRUNOL4 | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
Bruno (Drosophila) -like 4, RNA binding protein, BRUNOL-4, BRUNOL4Bruno -like 4, RNA binding protein, bruno-like 4, RNA binding protein, bruno-like 4, RNA binding protein (Drosophila), Bruno-like protein 4, CELF-4, CUG-BP and ETR-3 like factor 4, CUG-BP- and ETR-3-like factor 4, CUGBP Elav-like family member 4, CUGBP, Elav-like family member 4, LYST-interacting protein LIP9, RNA-binding protein BRUNOL4, RNA-binding protein BRUNOL-4 | |
CELF4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
0.2mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
56853 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MYIKMATLANGQADNASLSTNGLGSSPGSAGHMNGLSHSPGNPSTIPMKDH | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title