Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BSA Blocking Buffer, 3% in PBS, PROMO, Thermo Scientific Chemicals

Catalog No. J61655APPM
Click to view available options
Quantity:
250 mL
500 mL

BSA (bovine serum albumin) blocking buffer, 3% in PBS, is a blocking agent ideal for saturating excess protein-binding sites on membranes.

General Description

  • BSA blocking buffer contains a serum albumin protein isolated from a cow with a high affinity for proteins

Application

  • BSA (bovine serum albumin) blocking buffer is used as a blocking reagent ideal for saturating excess protein-binding sites on membranes and microplates for Western blotting and ELISA applications
  • This buffer can be used in a range from 0.3% to 5% dissolved in PBS solution
TRUSTED_SUSTAINABILITY

Specifications

Product Type Blocking Buffer
Quantity 500 mL
Form Liquid
Concentration 3% in PBS
Content And Storage -30°C to -10°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.