Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BST2 Antibody (2E2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP229621J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229621J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229621J Supplier Novus Biologicals Supplier No. NBP229621APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BST2 Monoclonal specifically detects BST2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BST2
Applications Flow Cytometry
Classification Monoclonal
Clone 200
Conjugate APC
Dilution Flow Cytometry
Gene Alias bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN
Gene Symbols BST2
Host Species Mouse
Immunogen Full length recombinant human BST2 protein [NCBI NP_004326]
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 684
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.