Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BST2 Antibody (44E9R), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB310668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310668 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310668 Supplier Novus Biologicals Supplier No. FAB8660PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

BST2 Monoclonal specifically detects BST2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen BST2
Applications Flow Cytometry
Classification Monoclonal
Clone 44E9R
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN
Host Species Rat
Immunogen Recombinant extracellular domain of CD317,
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 684
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.