Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BST2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227154SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227154SS 0.025 mg
NBP227154 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227154SS Supplier Novus Biologicals Supplier No. NBP227154SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

BST2 Polyclonal specifically detects BST2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Immunofluorescence.

Specifications

Antigen BST2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 4 ug/mL, Simple Western 1:200, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/mL, Immunohistochemistry-Frozen reported by customer review, Flow (Cell Surface) reported in scientific literature (PMID 21695198), Immunofluorescence
Formulation 0.1 ml PBS with 0.05% Sodium Azide
Gene Accession No. Q10589
Gene Alias bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN
Gene Symbols BST2
Host Species Rabbit
Immunogen A portion of amino acids 80-130 of human BST2 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 684
Test Specificity The BST2 amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, orangutan, white-cheeked gibbon, great ape, dwarf chimpanzee, gibbon, and 94% homologous in rhesus monkey.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.